Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxyquinoline-4-carboxylic acid, 98%

$79.72 - $207.93

Chemical Identifiers

Chemical Name or Material 2-Hydroxyquinoline-4-carboxylic acid
CAS 15733-89-8
Molecular Formula C10H7NO3
Formula Weight 189.17
MDL Number MFCD00023942
Synonym 1,2-Dihydro-2-oxoquinoline-4-carboxylic acid; 2-Hydroxycinchoninic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2924703

  SDS
alfa aesar™
H2924703
1g Each for $79.72

AAH2924706

  SDS
alfa aesar™
H2924706
5g Each for $207.93
Description & Specifications

Specifications

Formula Weight 189.17
Chemical Name or Material 2-Hydroxyquinoline-4-carboxylic acid
Assay 98%
Molecular Formula C10H7NO3
MDL Number MFCD00023942
CAS 15733-89-8
Synonym 1,2-Dihydro-2-oxoquinoline-4-carboxylic acid; 2-Hydroxycinchoninic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.