Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxypyrimidine hydrochloride, 98+%

$84.81 - $905.09

Chemical Identifiers

Chemical Name or Material 2-Hydroxypyrimidine hydrochloride
CAS 38353-09-2
Molecular Formula C4H4N2O·HCl
Formula Weight 132.55
MDL Number MFCD00012781
Beilstein 3560963
Synonym 2-Pyrimidinol hydrochloride; 2(1H)-Pyrimidinone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1373214

 
alfa aesar™
A1373214
25g Each for $84.81

AAA1373222

 
alfa aesar™
A1373222
100g Each for $221.66

AAA1373236

 
alfa aesar™
A1373236
500g Each for $905.09
Description & Specifications

Specifications

Formula Weight 132.55
Chemical Name or Material 2-Hydroxypyrimidine hydrochloride
TSCA TSCA
Assay ≥98%
Molecular Formula C4H4N2O·HCl
MDL Number MFCD00012781
CAS 38353-09-2
Beilstein 3560963
Synonym 2-Pyrimidinol hydrochloride; 2(1H)-Pyrimidinone hydrochloride
Melting Point ≈210°C (dec.)
Quantity 25g