missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxypyridine N-oxide, 98+%

Manufacturer:  Alfa Aesar™ H5420722

 View more versions of this product

Catalog No. AAH5420722



Specifications

Specifications

2-Hydroxypyridine N-oxide
100°C (212°F)
100g
C5H5NO2
1422545
SNUSZUYTMHKCPM-UHFFFAOYSA-N
1-hydroxypyridin-2-one
69975
≥98%
1.111
147°C to 152°C
13161-30-3
MFCD00006195
1-hydroxy-1,2-dihydropyridin-2-one
C1=CC(=O)N(C=C1)O
111.1
111.1

Chemical Identifiers

13161-30-3
111.1
SNUSZUYTMHKCPM-UHFFFAOYSA-N
69975
C1=CC(=O)N(C=C1)O
C5H5NO2
MFCD00006195
1-hydroxy-1,2-dihydropyridin-2-one
1-hydroxypyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-100-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents