missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxypyridine-4-carboxylic acid, 98%

$56.28 - $888.26

Chemical Identifiers

CAS 22282-72-0
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD01318566
InChI Key BXHCJLRTXPHUGH-UHFFFAOYSA-N
Synonym 2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid
PubChem CID 280756
IUPAC Name 2-oxo-1H-pyridine-4-carboxylic acid
SMILES C1=CNC(=O)C=C1C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6424106 View Documents Alfa Aesar™
H6424106
5g Each for $56.28
 
AAH6424114 View Documents Alfa Aesar™
H6424114
25g Each for $222.07
 
AAH6424122 View Documents Alfa Aesar™
H6424122
100g Each for $888.26
 
Description

Description

Applications
Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

22282-72-0
139.11
BXHCJLRTXPHUGH-UHFFFAOYSA-N
280756
C1=CNC(=O)C=C1C(=O)O
C6H5NO3
MFCD01318566
2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid
2-oxo-1H-pyridine-4-carboxylic acid

Specifications

5g
C6H5NO3
115859
BXHCJLRTXPHUGH-UHFFFAOYSA-N
2-oxo-1H-pyridine-4-carboxylic acid
280756
Solid
2-Hydroxypyridine-4-carboxylic acid
22282-72-0
MFCD01318566
2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid
C1=CNC(=O)C=C1C(=O)O
139.11
139.11
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning