Please login to your online account to display your discounted pricing

2'-Hydroxypropiophenone 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC121800050

 View more versions of this product

Catalog No. AC121800050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 150.18
Quantity 5g
Color Clear brown
TSCA TSCA
Density 1.0900g/mL
EINECS Number 210-244-1
Beilstein 08, 102
CAS Max % 100.0
CAS Min % 96.0
Refractive Index 1.5470 to 1.5490 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 610-99-1
Linear Formula HOC6H4COC2H5
Assay Percent Range 97%
Molecular Formula C9H10O2
Name Note 97%
MDL Number MFCD00002220
Chemical Name or Material 2';-Hydroxypropiophenone

Specifications

Chemical Name or Material 2';-Hydroxypropiophenone
Name Note 97%
CAS 610-99-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O2
Linear Formula HOC6H4COC2H5
Formula Weight 150.18
MDL Number MFCD00002220
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents