Please login to your online account to display your discounted pricing

2'-Hydroxypropiophenone 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2';-Hydroxypropiophenone
Name Note 97%
CAS 610-99-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O2
Linear Formula HOC6H4COC2H5
Formula Weight 150.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121805000

  SDS
thermo fisher scientific
121805000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC121801000

  SDS
fisher scientific acros
121801000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC121800050

  SDS
thermo fisher scientific
AC121800050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 150.18
Quantity 500g
Color Clear brown
Beilstein 08, 102
EINECS Number 210-244-1
TSCA TSCA
Density 1.0900g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5470 to 1.5490 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 2';-Hydroxypropiophenone
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H10O2
Linear Formula HOC6H4COC2H5
MDL Number MFCD00002220
CAS 610-99-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.