Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxynicotinic acid, 99%

$66.91 - $800.09

Chemical Identifiers

Chemical Name or Material 2-Hydroxynicotinic acid
CAS 609-71-2
Molecular Formula C6H5NO3
Formula Weight 139.11
MDL Number MFCD00010100
Beilstein 119028
Synonym 2-Hydroxypyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1598614

  SDS
alfa aesar™
A1598614
25g Each for $66.91

AAA1598622

  SDS
alfa aesar™
A1598622
100g Each for $198.89

AAA1598636

  SDS
alfa aesar™
A1598636
500g Each for $800.09
Description & Specifications

Specifications

Formula Weight 139.11
Synonym 2-Hydroxypyridine-3-carboxylic acid
Beilstein 119028
Quantity 25g
Molecular Formula C6H5NO3
CAS 609-71-2
MDL Number MFCD00010100
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 256° to 261°C
Assay 99%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Hydroxynicotinic acid

Applications
2-Hydroxynicotinic acid is used in chemical synthesis. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in sodium hydroxide (0.1 g/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.