missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxymethylmorpholine hydrochloride, 95%

$269.66 - $805.66

Chemical Identifiers

CAS 144053-98-5
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.606
MDL Number MFCD13191721
InChI Key NBGXGDBTUJNTKJ-UHFFFAOYSA-N
Synonym 2-hydroxymethyl-morpholine hydrochloride, 2-morpholinemethanol hcl, 2-morpholinemethanol hydrochloride, morpholin-2-yl methanol hcl, morpholin-2-yl methanol hydrochloride, morpholin-2-yl-methanol hydrochloride, morpholin-2-ylmethanol hcl, morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethyl morpholine hydrochloride, s-2-hydroxymethyl morpholine hydrochloride
PubChem CID 19777263
IUPAC Name morpholin-2-ylmethanol;hydrochloride
SMILES C1COC(CN1)CO.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64154MD View Documents Alfa Aesar™
H64154MD
250mg Each for $269.66
 
AAH6415403 View Documents Alfa Aesar™
H6415403
1g Each for $805.66
 
Specifications

Chemical Identifiers

144053-98-5
153.606
NBGXGDBTUJNTKJ-UHFFFAOYSA-N
19777263
C1COC(CN1)CO.Cl
C5H12ClNO2
MFCD13191721
2-hydroxymethyl-morpholine hydrochloride, 2-morpholinemethanol hcl, 2-morpholinemethanol hydrochloride, morpholin-2-yl methanol hcl, morpholin-2-yl methanol hydrochloride, morpholin-2-yl-methanol hydrochloride, morpholin-2-ylmethanol hcl, morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethyl morpholine hydrochloride, s-2-hydroxymethyl morpholine hydrochloride
morpholin-2-ylmethanol;hydrochloride

Specifications

250mg
C5H12ClNO2
7301430
NBGXGDBTUJNTKJ-UHFFFAOYSA-N
morpholin-2-ylmethanol;hydrochloride
19777263
Solid
2-Hydroxymethylmorpholine hydrochloride
144053-98-5
MFCD13191721
2-hydroxymethyl-morpholine hydrochloride, 2-morpholinemethanol hcl, 2-morpholinemethanol hydrochloride, morpholin-2-yl methanol hcl, morpholin-2-yl methanol hydrochloride, morpholin-2-yl-methanol hydrochloride, morpholin-2-ylmethanol hcl, morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethyl morpholine hydrochloride, s-2-hydroxymethyl morpholine hydrochloride
C1COC(CN1)CO.Cl
153.606
153.61
95%
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures