missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxymethylmorpholine hydrochloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6415403
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Hydroxymethylmorpholine hydrochloride | |
C5H12ClNO2 | |
1 g | |
morpholin-2-ylmethanol hydrochloride, 2-morpholinemethanol hcl, 2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride, s-2-hydroxymethyl morpholine hydrochloride, morpholin-2-yl methanol hcl, 2-morpholinemethanol hydrochloride, morpholin-2-ylmethanol hcl, morpholin-2-yl-methanol hydrochloride, morpholin-2-yl methanol hydrochloride | |
C1COC(CN1)CO.Cl | |
153.606 | |
153.61 | |
Solid |
144053-98-5 | |
MFCD13191721 | |
7301430 | |
NBGXGDBTUJNTKJ-UHFFFAOYSA-N | |
morpholin-2-ylmethanol;hydrochloride | |
19777263 | |
95% |
Chemical Identifiers
144053-98-5 | |
153.606 | |
NBGXGDBTUJNTKJ-UHFFFAOYSA-N | |
19777263 | |
C1COC(CN1)CO.Cl |
C5H12ClNO2 | |
MFCD13191721 | |
morpholin-2-ylmethanol hydrochloride, 2-morpholinemethanol hcl, 2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride, s-2-hydroxymethyl morpholine hydrochloride, morpholin-2-yl methanol hcl, 2-morpholinemethanol hydrochloride, morpholin-2-ylmethanol hcl, morpholin-2-yl-methanol hydrochloride, morpholin-2-yl methanol hydrochloride | |
morpholin-2-ylmethanol;hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures