Please login to your online account to display your discounted pricing

2-(Hydroxymethyl)anthraquinone, 97%, Acros Organics

$278.28 - $278.28

Chemical Identifiers

Chemical Name or Material 2-(Hydroxymethyl)anthraquinone
Name Note 97%
CAS 17241-59-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C15H10O3
Formula Weight 238.24
MDL Number MFCD00001236
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC210200050

  SDS
thermo fisher scientific
210200050
5g Each for $278.28
Description & Specifications

Specifications

Chemical Name or Material 2-(Hydroxymethyl)anthraquinone
Color Yellow to brown
Melting Point 192° to 197°C
Quantity 5g
Name Note 97%
CAS 17241-59-7
CAS Min % 96.0
Health Hazard 1 LIGHT SENSITIVE
CAS Max % 100.0
RTECS Number CB7600000
EINECS Number 241-274-3
TSCA TSCA
Infrared Spectrum Authentic
Physical Form Crystalline Powder
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C15H10O3
Formula Weight 238.24
MDL Number MFCD00001236
Beilstein 08, III, 2940
Fieser 08,252; 02,369; 04,432; 05,585; 06,518; 15,119; 16,120; 17,105