missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxymethyl-5-nitrobenzeneboronic acid hemiester, 96%

Manufacturer:  Alfa Aesar™ H53212MD

 View more versions of this product

Catalog No. AAH53212MD



Specifications

Specifications

2-Hydroxymethyl-5-nitrobenzeneboronic acid hemiester
118803-40-0
MFCD04115657
GFFKBQCIGADRSN-UHFFFAOYSA-N
1-hydroxy-6-nitro-3H-2,1-benzoxaborole
3813634
96%
250mg
C7H6BNO4
6-nitrobenzo c 1,2 oxaborol-1 3h-ol, 1-hydroxy-6-nitro-2,1-benzoxaborolane, 2-hydroxymethyl-5-nitro benzeneboronic acid dehydrate, 6-nitro-3h-2,1-benzoxaborol-1-ol, 6-nitro-1,3-dihydro-2,1-benzoxaborol-1-ol, 2-hydroxymethyl-5-nitro phenylboronic acid, dehydrate, 2-hydroxymethyl-5-nitrophenylboronic acid, dehydrated, 6-nitro-2,1-benzoxaborol-1 3h-ol, 1,3-dihydro-1-hydroxy-6-nitro-2,1-benzoxaborole, 2,1-benzoxaborole,1,3-dihydro-1-hydroxy-6-nitro
B1(C2=C(CO1)C=CC(=C2)[N+](=O)[O-])O
178.938
178.94

Chemical Identifiers

118803-40-0
178.938
GFFKBQCIGADRSN-UHFFFAOYSA-N
3813634
B1(C2=C(CO1)C=CC(=C2)[N+](=O)[O-])O
C7H6BNO4
MFCD04115657
6-nitrobenzo c 1,2 oxaborol-1 3h-ol, 1-hydroxy-6-nitro-2,1-benzoxaborolane, 2-hydroxymethyl-5-nitro benzeneboronic acid dehydrate, 6-nitro-3h-2,1-benzoxaborol-1-ol, 6-nitro-1,3-dihydro-2,1-benzoxaborol-1-ol, 2-hydroxymethyl-5-nitro phenylboronic acid, dehydrate, 2-hydroxymethyl-5-nitrophenylboronic acid, dehydrated, 6-nitro-2,1-benzoxaborol-1 3h-ol, 1,3-dihydro-1-hydroxy-6-nitro-2,1-benzoxaborole, 2,1-benzoxaborole,1,3-dihydro-1-hydroxy-6-nitro
1-hydroxy-6-nitro-3H-2,1-benzoxaborole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents