missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Hydroxymethyl)-18-crown 6-Ether 93.0+%, TCI America™
Supplier: TCI America H09901G
Specifications
| 2-(Hydroxymethyl)-18-crown 6-Ether | |
| C13H26O7 | |
| 1 g | |
| HFRGASADQCZXHH-UHFFFAOYNA-N | |
| (1,4,7,10,13,16-hexaoxacyclooctadecan-2-yl)methanol | |
| 3611393 | |
| ≥93.0% (GC) |
| 70069-04-4 | |
| MFCD00188046 | |
| 1,4,7,10,13,16-hexaoxacyclooctadecane-2-methanol, 2-hydroxymethyl-18-crown-6, 2-hydroxymethyl-18-crown 6-ether, 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol, hydroxy-18-crown-6, 18-crown-6-methanol, hydroxymethyl-18-crown-6, hfrgasadqczxhh-uhfffaoysa, 2-hydroxymethyl-18-crown, 2,5,8,11,14,17-hexaoxacyclooctadecyl methan-1-ol | |
| OCC1COCCOCCOCCOCCOCCO1 | |
| 294.34 | |
| 294.34 | |
| Liquid |
Chemical Identifiers
| 70069-04-4 | |
| 294.34 | |
| HFRGASADQCZXHH-UHFFFAOYNA-N | |
| 3611393 | |
| OCC1COCCOCCOCCOCCOCCO1 |
| C13H26O7 | |
| MFCD00188046 | |
| 1,4,7,10,13,16-hexaoxacyclooctadecane-2-methanol, 2-hydroxymethyl-18-crown-6, 2-hydroxymethyl-18-crown 6-ether, 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol, hydroxy-18-crown-6, 18-crown-6-methanol, hydroxymethyl-18-crown-6, hfrgasadqczxhh-uhfffaoysa, 2-hydroxymethyl-18-crown, 2,5,8,11,14,17-hexaoxacyclooctadecyl methan-1-ol | |
| (1,4,7,10,13,16-hexaoxacyclooctadecan-2-yl)methanol |
Safety and Handling
TSCA : No