missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxymethyl-18-crown-6, 97%

Manufacturer:  Alfa Aesar™ 4447701

 View more versions of this product

Catalog No. AA4447701



Specifications

Specifications

2-Hydroxymethyl-18-crown-6
>110°C (230°F)
150°C (0.5mmHg)
70069-04-4
MFCD00188046
1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol
HFRGASADQCZXHH-UHFFFAOYSA-N
1,4,7,10,13,16-hexaoxacyclooctadec-2-ylmethanol
3611393
Pale yellow liquid
1.174 g/mL
1.479
0.25g
C13H26O7
4313301
Soluble in water (1000g/L) (25°C).
C1COCCOCCOC(COCCOCCO1)CO
294.344
294.35
97%

Chemical Identifiers

70069-04-4
294.344
HFRGASADQCZXHH-UHFFFAOYSA-N
3611393
C1COCCOCCOC(COCCOCCO1)CO
C13H26O7
MFCD00188046
1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol
1,4,7,10,13,16-hexaoxacyclooctadec-2-ylmethanol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 274-299-3

TSCA : No

Recommended Storage : Ambient temperatures

Documents