Please login to your online account to display your discounted pricing

2-Hydroxymethyl-15-crown-5, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Hydroxymethyl-15-crown-5
CAS 75507-25-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C11H22O6
Formula Weight 250.29
MDL Number MFCD00192215
Synonym 2-(Hydroxymethyl)-1,4,7,10,13-pentaoxacyclopentadecane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336250010

  SDS
thermo fisher scientific
336250010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 250.29
Quantity 1g
Color Clear colorless to almost colorless
Density 1.1g/mL
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2-Hydroxymethyl-15-crown-5
Refractive Index 1.4780 to 1.4800 (20°C, 589nm)
CAS 75507-25-4
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C11H22O6
MDL Number MFCD00192215
Synonym 2-(Hydroxymethyl)-1,4,7,10,13-pentaoxacyclopentadecane
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Chelation/Complexation reagent

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.