missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxymethyl-12-crown-4, 97%

Manufacturer:  Alfa Aesar™ 4450901

 View more versions of this product

Catalog No. AA4450901



Specifications

Specifications

2-Hydroxymethyl-12-crown-4
>110°C (230°F)
115°C (0.04mmHg)
75507-26-5
MFCD00067524
1,4,7,10-tetraoxacyclododecan-2-methanol
NJIPEIQHUNDGPY-UHFFFAOYSA-N
1,4,7,10-tetraoxacyclododec-2-ylmethanol
2733085
Liquid
1.20 g/mL
1.48
0.25g
C9H18O5
4661968
Miscible in water. Soluble in non-polar solvents. Insoluble in water.
C1COCCOC(COCCO1)CO
206.238
206.24
97%

Chemical Identifiers

75507-26-5
206.238
NJIPEIQHUNDGPY-UHFFFAOYSA-N
2733085
C1COCCOC(COCCO1)CO
C9H18O5
MFCD00067524
1,4,7,10-tetraoxacyclododecan-2-methanol
1,4,7,10-tetraoxacyclododec-2-ylmethanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 278-224-5

TSCA : No

Recommended Storage : Ambient temperatures

Documents