missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyisophthalaldehyde, TCI America™

$491.95 - $1,865.45

Chemical Identifiers

CAS 3328-69-6
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00839420
InChI Key JJOPQMAMJLOGFB-UHFFFAOYSA-N
Synonym 2,6-Diformylphenol
PubChem CID 345554
IUPAC Name 2-hydroxybenzene-1,3-dicarbaldehyde
SMILES C1=CC(=C(C(=C1)C=O)O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10181G View Documents TCI America
H10181G
1g Each for $491.95
 
H10185G View Documents TCI America
H10185G
5g Each for $1,865.45
 
Specifications

Chemical Identifiers

3328-69-6
150.133
JJOPQMAMJLOGFB-UHFFFAOYSA-N
345554
C1=CC(=C(C(=C1)C=O)O)C=O
C8H6O3
MFCD00839420
2,6-Diformylphenol
2-hydroxybenzene-1,3-dicarbaldehyde

Specifications

Undesignated
1g
C8H6O3
2,6-Diformylphenol
C1=CC(=C(C(=C1)C=O)O)C=O
150.133
150.13
≥98.0% (GC,T)
124°C
3328-69-6
MFCD00839420
JJOPQMAMJLOGFB-UHFFFAOYSA-N
2-hydroxybenzene-1,3-dicarbaldehyde
345554
Crystal-Powder at 20°C
2-Hydroxyisophthalaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator