Learn More
2-Hydroxyisobutyric acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 173100250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Hydroxyisobutyric acid | |
594-61-6 | |
100.0 | |
98% | |
C4H8O3 | |
MFCD00004459 | |
03, 313 | |
Solubility in water: 1900g/L (22°C). Other solubilities: soluble in methanol,diethyl ether and hot benzene,sparingly soluble in cold benzene | |
CC(C)(O)C(O)=O | |
104.11 | |
CHEBI:50129 | |
98% |
98% | |
97.5 | |
84°C (1.5mmHg) | |
Plastic bottle | |
(CH3)2C(OH)CO2H | |
25g | |
2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid | |
BWLBGMIXKSTLSX-UHFFFAOYSA-N | |
2-hydroxy-2-methylpropanoic acid | |
11671 | |
104.11 |
Chemical Identifiers
594-61-6 | |
104.11 | |
BWLBGMIXKSTLSX-UHFFFAOYSA-N | |
11671 | |
2-hydroxy-2-methylpropanoic acid |
C4H8O3 | |
MFCD00004459 | |
2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid | |
CHEBI:50129 | |
CC(C)(O)C(O)=O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 209-848-8
TSCA : TSCA
RUO â Research Use Only