Please login to your online account to display your discounted pricing

2-Hydroxyethyledisulfide, 90%, ACROS Organics™

$55.18 - $55.18

Chemical Identifiers

Chemical Name or Material 2-Hydroxyethyledisulfide
CAS 1892-29-1
CAS Min % 88.0
CAS Max % 100.0
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C4H10O2S2
Linear Formula (HOCH2CH2)2S2
Formula Weight 154.25
MDL Number MFCD00002906
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC373895000

  SDS
thermo fisher scientific
AC373895000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC373890250

  SDS
thermo fisher scientific
AC373890250
Glass bottle 25g Each for $55.18

AC373891000

  SDS
thermo fisher scientific
AC373891000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88%
Physical Form Viscous Liquid
Formula Weight 154.25
Quantity 500g
Color Clear colorless to yellow
Density 1.2877g/mL
Beilstein 01, 471
CAS Min % 88.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
Chemical Name or Material 2-Hydroxyethyledisulfide
Infrared Spectrum Authentic
Molecular Formula C4H10O2S2
Assay Percent Range 90%
CAS 1892-29-1
MDL Number MFCD00002906
Linear Formula (HOCH2CH2)2S2
Synonym 2, 2'-Dithiodiethanol
Water 10% max. (K.F.)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.