Please login to your online account to display your discounted pricing

2-Hydroxyethyledisulfide, 90%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC373891000

 View more versions of this product

Catalog No. AC373891000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥88%
Physical Form Viscous Liquid
Formula Weight 154.25
Quantity 100g
Color Clear colorless to yellow
Beilstein 01, 471
Density 1.2877g/mL
CAS Max % 100.0
CAS Min % 88.0
Chemical Name or Material 2-Hydroxyethyledisulfide
Water 10% max. (K.F.)
Molecular Formula C4H10O2S2
Synonym 2, 2'-Dithiodiethanol
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1892-29-1
Assay Percent Range 90%
MDL Number MFCD00002906
Packaging Glass bottle
Linear Formula (HOCH2CH2)2S2
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Hydroxyethyledisulfide
CAS 1892-29-1
CAS Min % 88.0
CAS Max % 100.0
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C4H10O2S2
Linear Formula (HOCH2CH2)2S2
Formula Weight 154.25
MDL Number MFCD00002906
Synonym 2, 2'-Dithiodiethanol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.

SDS & Safety Documents