Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Hydroxyethyl)trimethylammonium dimethyl phosphate, 95%

$80.19 - $378.66

Chemical Identifiers

Chemical Name or Material (2-Hydroxyethyl)trimethylammonium dimethyl phosphate
CAS 118978-98-6
Molecular Formula C7H20NO5P
Linear Formula (HOCH2CH2)N(CH3)3·(CH3O)2PO2
Formula Weight 229.21
MDL Number MFCD09953490
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2760906

  SDS
alfa aesar™
H2760906
5g Each for $80.19

AAH2760918

  SDS
alfa aesar™
H2760918
50g Each for $378.66
Description & Specifications

Specifications

Formula Weight 229.21
Chemical Name or Material (2-Hydroxyethyl)trimethylammonium dimethyl phosphate
Assay 95%
Molecular Formula C7H20NO5P
Linear Formula (HOCH2CH2)N(CH3)3·(CH3O)2PO2
MDL Number MFCD09953490
CAS 118978-98-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Melting Point 40° to 41°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.