missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxyethyl stearate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 346951000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Hydroxyethyl stearate | |
55°C to 60°C | |
149°C | |
Glass bottle | |
MFCD00051465 | |
glycol stearate, cremophor a, 2-hydroxyethyl stearate, polyethylene glycol monostearate, ethylene glycol monostearate, cerasynt m, clearate g, cerasynt mn, cithrol ps, polyoxyl 40 stearate | |
RFVNOJDQRGSOEL-UHFFFAOYSA-N | |
2-hydroxyethyl octadecanoate | |
24762 | |
Technical | |
Flakes |
111-60-4 | |
White to Yellow | |
Authentic | |
C20H40O3 | |
100 g | |
Solubility in water: insoluble in h2o | |
CCCCCCCCCCCCCCCCCC(=O)OCCO | |
328.53 | |
328.53 | |
compound is a mixture of various chain-length esters and di(ethylene glycol) ester |
Chemical Identifiers
111-60-4 | |
328.53 | |
RFVNOJDQRGSOEL-UHFFFAOYSA-N | |
24762 | |
CCCCCCCCCCCCCCCCCC(=O)OCCO |
C20H40O3 | |
MFCD00051465 | |
glycol stearate, cremophor a, 2-hydroxyethyl stearate, polyethylene glycol monostearate, ethylene glycol monostearate, cerasynt m, clearate g, cerasynt mn, cithrol ps, polyoxyl 40 stearate | |
2-hydroxyethyl octadecanoate |
Safety and Handling
EINECSNumber : 203-886-9
RTECSNumber : WI4100000
TSCA : TSCA
RUO – Research Use Only