missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyethyl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™

Manufacturer:  TCI America M0085500G

 View more versions of this product

Catalog No. M0085500G



Specifications

Specifications

2-Hydroxyethyl Methacrylate (stabilized with MEHQ)
Yellow
868-77-9
MFCD00002863
WOBHKFSMXKNTIM-UHFFFAOYSA-N
2-hydroxyethyl 2-methylprop-2-enoate
13360
130.14
≥95.0% (GC)
85°C
500g
C6H10O3
2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
CC(=C)C(=O)OCCO
130.143
CHEBI:34288
Clear Liquid at 20°C

Chemical Identifiers

868-77-9
130.143
WOBHKFSMXKNTIM-UHFFFAOYSA-N
13360
2-hydroxyethyl 2-methylprop-2-enoate
C6H10O3
MFCD00002863
2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
CHEBI:34288
CC(=C)C(=O)OCCO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0958&(2)-1044

RTECSNumber : OZ4725000

TSCA : Yes

Documents