missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyethyl methacrylate 97%, stabilized, ACROS Organics™

Manufacturer:  Acros Organics 156330250

 View more versions of this product

Catalog No. AC156330250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2mg KOH/g max.
1.073g/mL
Glass bottle
200 to 400ppm MEHQ
Undesignated
868-77-9,150-76-5
100.0
C6H10O3
MFCD00002863
2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
CC(=C)C(=O)OCCO
130.143
CHEBI:34288
Liquid
2-Hydroxyethyl methacrylate
Authentic
1.4510 to 1.4530 (20°C, 589nm)
0.2% max. (K.F.)
25g
96.0
97%
H2C=C(CH3)CO2CH2CH2OH
02, IV, 1530
WOBHKFSMXKNTIM-UHFFFAOYSA-N
2-hydroxyethyl 2-methylprop-2-enoate
13360
130.14
≥96.0%

Chemical Identifiers

868-77-9
130.143
WOBHKFSMXKNTIM-UHFFFAOYSA-N
13360
2-hydroxyethyl 2-methylprop-2-enoate
C6H10O3
MFCD00002863
2-hydroxyethyl methacrylate, glycol methacrylate, hydroxyethyl methacrylate, glycol monomethacrylate, hema, ethylene glycol methacrylate, 2-methacryloyloxy ethanol, 2-hydroxyethylmethacrylate, mhoromer, monomer mg-1
CHEBI:34288
CC(=C)C(=O)OCCO
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents