missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04379309

 View more versions of this product

Catalog No. AA4379309



Description

Description

(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(2-Hydroxyethyl)hydrazine
-70°C
218°C to 220°C
C2H8N2O
MFCD00007623
UN2810
Soluble in water,alcohols and glycols.
C(CO)NN
76.099
CHEBI:50098
Liquid
109-84-2
1.121 g/mL
73°C (163°F)
1.493
10 g
2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
GBHCABUWWQUMAJ-UHFFFAOYSA-N
2-hydrazinylethanol
8017
76.1

Chemical Identifiers

109-84-2
76.099
GBHCABUWWQUMAJ-UHFFFAOYSA-N
8017
2-hydrazinylethanol
C2H8N2O
MFCD00007623
2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
CHEBI:50098
C(CO)NN
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H301-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 203-711-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only