missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyethyl acrylate, 97%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 334080050

 View more versions of this product

Catalog No. AC334080050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0% (GC)
Oily Liquid
Yellow to brown
96.0
1.4470 to 1.4520 (20°C, 589nm)
200 to 450ppm MEHQ
97%
150-76-5,818-61-1
diester (GC): < 0.2%
0.15% max.
C5H8O3
116.12
5mL
1.1060g/mL
100.0
Authentic
Stabilized
2-Hydroxyethyl acrylate
4mg KOH/g max.
Glass bottle
MFCD00002865

Chemical Identifiers

818-61-1,150-76-5
100.0
C5H8O3
MFCD00002865
≥96.0% (GC)
96.0
97%
116.12
2-Hydroxyethyl acrylate, Stabilized

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger