missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxydiphenylmethane, 98%, ACROS Organics™

Manufacturer:  Acros Organics 162000050

 View more versions of this product

Catalog No. AC162000050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Hydroxydiphenylmethane
Glass Bottle
51°C to 55°C
98%
97.5
98%
C6H5CH2C6H4OH
2-hydroxydiphenylmethane, o-benzylphenol, phenol, 2-phenylmethyl, benzylphenol, 2-benzyl-phenol, 2-phenylmethyl phenol, alpha-phenyl-o-cresol, o-cresol, .alpha.-phenyl, unii-sf555bl24o, 2-benzyl phenol
C1=CC=C(C=C1)CC2=CC=CC=C2O
184.238
184.24
≥97.5%
Authentic
Colorless to Pink-Red
5g
28994-41-4
100.0
C13H12O
MFCD00002242
CDMGNVWZXRKJNS-UHFFFAOYSA-N
2-benzylphenol
24216
Crystals

Chemical Identifiers

28994-41-4
184.238
CDMGNVWZXRKJNS-UHFFFAOYSA-N
24216
C1=CC=C(C=C1)CC2=CC=CC=C2O
C13H12O
MFCD00002242
2-hydroxydiphenylmethane, o-benzylphenol, phenol, 2-phenylmethyl, benzylphenol, 2-benzyl-phenol, 2-phenylmethyl phenol, alpha-phenyl-o-cresol, o-cresol, .alpha.-phenyl, unii-sf555bl24o, 2-benzyl phenol
2-benzylphenol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 249-361-8

TSCA : TSCA

SDS Documents