missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxydiphenylmethane, 98%, ACROS Organics™

Chemical Identifiers

CAS 28994-41-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C13H12O
Linear Formula C6H5CH2C6H4OH
Formula Weight 184.24
MDL Number MFCD00002242
Synonym α-Phenyl-o-cresol
Chemical Name or Material 2-Hydroxydiphenylmethane, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC162000050 SDS Thermo Fisher Scientific
162000050
Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

28994-41-4
100.0
C13H12O
184.24
α-Phenyl-o-cresol
≥97.5%
97.5
98%
C6H5CH2C6H4OH
MFCD00002242
2-Hydroxydiphenylmethane, 98%

Specifications

Crystals
5g
Glass Bottle
100.0
α-Phenyl-o-cresol
MFCD00002242
98%
28994-41-4
≥97.5%
184.24
Colorless to red-pink
51° to 55°C
97.5
Authentic
C13H12O
C6H5CH2C6H4OH
2-Hydroxydiphenylmethane, 98%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 249-361-8

TSCA : TSCA