missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxycyclohexanone, dimer +90%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393051000

 View more versions of this product

Catalog No. AC393051000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Hydroxycyclohexanone, dimer
100g
90.0
Authentic
Powder
90+%
228.29
Adipoin
Off-white
30282-14-5
100.0
Glass bottle
≥90.0%
C12H20O4
MFCD00151157

Chemical Identifiers

30282-14-5
100.0
C12H20O4
MFCD00151157
2-Hydroxycyclohexanone, dimer
90.0
90+%
228.29
Adipoin
≥90.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning