Learn More
2-Hydroxycarbazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2776506
Description
Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCarbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2-Hydroxycarbazole | |
262°C to 273°C | |
C12H9NO | |
5 g | |
GWPGDZPXOZATKL-UHFFFAOYSA-N | |
9H-carbazol-2-ol | |
93551 | |
97% |
86-79-3 | |
Odorless | |
MFCD00004962 | |
2-hydroxycarbazole, carbazol-2-ol, ccris 5301, 2-hydroxycarbazol, 7-hydroxycarbazole, 2-hydroxy carbazole, pubchem9945, pubchem9949, 2-hydroxy-9h-carbazol, 2-hydroxy-9h-carbazole | |
OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 | |
183.21 | |
183.21 |
Chemical Identifiers
86-79-3 | |
183.21 | |
GWPGDZPXOZATKL-UHFFFAOYSA-N | |
93551 | |
OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1 |
C12H9NO | |
MFCD00004962 | |
2-hydroxycarbazole, carbazol-2-ol, ccris 5301, 2-hydroxycarbazol, 7-hydroxycarbazole, 2-hydroxy carbazole, pubchem9945, pubchem9949, 2-hydroxy-9h-carbazol, 2-hydroxy-9h-carbazole | |
9H-carbazol-2-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-699-7
RTECSNumber : FE6355000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only