missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Hydroxybenzyl)triphenylphosphonium Bromide, TCI America™

$41.25 - $154.31

Chemical Identifiers

CAS 70340-04-4
Molecular Formula C25H22BrOP
Molecular Weight (g/mol) 449.328
MDL Number MFCD00011902
InChI Key FKKZGQXMWVGPMH-UHFFFAOYSA-N
Synonym 2-hydroxybenzyl triphenylphosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphanium bromide, 2-hydroxyphenyl methyl triphenylphosphanium bromide, phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide, acmc-209oe4, 2-hydroxybenzyltriphenylphosphonium bromide, 2-hydroxybenzyl triphenylphosphoniumbromide, 2-hydroxybenzyl triphenyl phosphonium bromide, ortho hydroxybenzyltriphenyl-phosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphonium bromide
PubChem CID 2724142
IUPAC Name (2-hydroxyphenyl)methyl-triphenylphosphanium;bromide
SMILES C1=CC=C(C=C1)[P+](CC2=CC=CC=C2O)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H124025G View Documents TCI America
H124025G
25g Each for $154.31
 
H12405G View Documents TCI America
H12405G
5g Each for $41.25
 
Specifications

Chemical Identifiers

70340-04-4
449.328
FKKZGQXMWVGPMH-UHFFFAOYSA-N
2724142
C1=CC=C(C=C1)[P+](CC2=CC=CC=C2O)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
C25H22BrOP
MFCD00011902
2-hydroxybenzyl triphenylphosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphanium bromide, 2-hydroxyphenyl methyl triphenylphosphanium bromide, phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide, acmc-209oe4, 2-hydroxybenzyltriphenylphosphonium bromide, 2-hydroxybenzyl triphenylphosphoniumbromide, 2-hydroxybenzyl triphenyl phosphonium bromide, ortho hydroxybenzyltriphenyl-phosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphonium bromide
(2-hydroxyphenyl)methyl-triphenylphosphanium;bromide

Specifications

Undesignated
25g
C25H22BrOP
2-hydroxybenzyl triphenylphosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphanium bromide, 2-hydroxyphenyl methyl triphenylphosphanium bromide, phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide, acmc-209oe4, 2-hydroxybenzyltriphenylphosphonium bromide, 2-hydroxybenzyl triphenylphosphoniumbromide, 2-hydroxybenzyl triphenyl phosphonium bromide, ortho hydroxybenzyltriphenyl-phosphonium bromide, 2-hydroxyphenyl methyl-triphenylphosphonium bromide
C1=CC=C(C=C1)[P+](CC2=CC=CC=C2O)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
449.328
449.33
≥97.0% (HPLC,T)
243°C
70340-04-4
MFCD00011902
FKKZGQXMWVGPMH-UHFFFAOYSA-N
(2-hydroxyphenyl)methyl-triphenylphosphanium;bromide
2724142
Crystal-Powder at 20°C
(2-Hydroxybenzyl)triphenylphosphonium Bromide
Safety and Handling

Safety and Handling

TSCA : No