missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Hydroxybenzyl)triphenylphosphonium bromide, 98%, ACROS Organics™

$35.98 - $35.98

Chemical Identifiers

CAS 70340-04-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C25H22BrOP
Linear Formula HOC6H4CH2P(C6H5)3Br
Formula Weight 449.33
MDL Number MFCD00011902
Chemical Name or Material (2-Hydroxybenzyl)triphenylphosphonium bromide, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC202070100 SDS Thermo Fisher Scientific
202070100
Glass bottle 10g Each for $35.98
Specifications

Description

For C-C Bond Formation (Olefination)
Description & Specifications

Chemical Identifiers

70340-04-4
100.0
C25H22BrOP
449.33
(2-Hydroxybenzyl)triphenylphosphonium bromide, 98%
97.5
98%
HOC6H4CH2P(C6H5)3Br
MFCD00011902
≥97.5%

Specifications

Powder
10g
240° to 244°C
97.5
Glass bottle
MFCD00011902
98%
(2-Hydroxybenzyl)triphenylphosphonium bromide, 98%
449.33
Off-white to light yellow
100.0
Authentic
70340-04-4
C25H22BrOP
HOC6H4CH2P(C6H5)3Br
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 274-574-8

TSCA : TSCA