missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Hydroxybenzyl)triphenylphosphonium bromide, 98+%

Manufacturer:  Alfa Aesar™ L0077809

 View more versions of this product

Catalog No. AAL0077809



Specifications

Specifications

Hygroscopic
10g
C25H22BrOP
4173355
FKKZGQXMWVGPMH-UHFFFAOYSA-N
(2-hydroxyphenyl)methyl-triphenylphosphanium;bromide
2724142
≥98%
240°C to 245°C
70340-04-4
MFCD00011902
2-hydroxybenzyl triphenyl phosphonium bromide, 2-hydroxybenzyl triphenylphosphonium bromide, 2-hydroxybenzyl triphenylphosphoniumbromide, 2-hydroxybenzyltriphenylphosphonium bromide, 2-hydroxyphenyl methyl triphenylphosphanium bromide, 2-hydroxyphenyl methyl-triphenylphosphanium bromide, 2-hydroxyphenyl methyl-triphenylphosphonium bromide, acmc-209oe4, ortho hydroxybenzyltriphenyl-phosphonium bromide, phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide
C1=CC=C(C=C1)[P+](CC2=CC=CC=C2O)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
449.328
449.34

Chemical Identifiers

70340-04-4
449.328
FKKZGQXMWVGPMH-UHFFFAOYSA-N
2724142
C1=CC=C(C=C1)[P+](CC2=CC=CC=C2O)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
C25H22BrOP
MFCD00011902
2-hydroxybenzyl triphenyl phosphonium bromide, 2-hydroxybenzyl triphenylphosphonium bromide, 2-hydroxybenzyl triphenylphosphoniumbromide, 2-hydroxybenzyltriphenylphosphonium bromide, 2-hydroxyphenyl methyl triphenylphosphanium bromide, 2-hydroxyphenyl methyl-triphenylphosphanium bromide, 2-hydroxyphenyl methyl-triphenylphosphonium bromide, acmc-209oe4, ortho hydroxybenzyltriphenyl-phosphonium bromide, phosphonium, 2-hydroxyphenyl methyl triphenyl-, bromide
(2-hydroxyphenyl)methyl-triphenylphosphanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 274-574-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents