Please login to your online account to display your discounted pricing

2-Hydroxybenzyl alcohol, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  173330250

 View more versions of this product

Catalog No. AC173330250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 124.14
Assay ≥96.0%
Physical Form Crystalline Flakes or Powder
Quantity 25g
Color Light beige
TSCA TSCA
RTECS Number DO6430000
EINECS Number 201-960-5
Melting Point 83° to 87°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym Salicyl alcohol
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Warning
Chemical Name or Material 2-Hydroxybenzyl alcohol
Molecular Formula C7H8O2
MDL Number MFCD00004617
Linear Formula HOC6H4CH2OH
CAS 90-01-7
Assay Percent Range 97%
Name Note 97%

Specifications

Chemical Name or Material 2-Hydroxybenzyl alcohol
Name Note 97%
CAS 90-01-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H8O2
Linear Formula HOC6H4CH2OH
Formula Weight 124.14
MDL Number MFCD00004617
Synonym Salicyl alcohol
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents