missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyanthraquinone, TCI America™

$151.25 - $750.16

Chemical Identifiers

CAS 605-32-3
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.215
MDL Number MFCD00027374
InChI Key GCDBEYOJCZLKMC-UHFFFAOYSA-N
PubChem CID 11796
ChEBI CHEBI:37482
IUPAC Name 2-hydroxyanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H101225G View Documents TCI America
H101225G
25g Each for $151.25
 
H1012250G View Documents TCI America
H1012250G
250g Each for $750.16
 
Specifications

Chemical Identifiers

605-32-3
224.215
GCDBEYOJCZLKMC-UHFFFAOYSA-N
CHEBI:37482
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)O
C14H8O3
MFCD00027374
11796
2-hydroxyanthracene-9,10-dione

Specifications

Undesignated
605-32-3
MFCD00027374
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)O
224.215
CHEBI:37482
Crystal-Powder at 20°C
2-Hydroxyanthraquinone
25g
C14H8O3
GCDBEYOJCZLKMC-UHFFFAOYSA-N
2-hydroxyanthracene-9,10-dione
11796
224.22
≥97.0% (HPLC,T)
Safety and Handling

Safety and Handling

RTECSNumber : CB7250000

TSCA : Yes