Please login to your online account to display your discounted pricing

2'-Hydroxyacetophenone 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2'-Hydroxyacetophenone
Name Note 99%
CAS 118-93-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H8O2
Linear Formula HOC6H4COCH3
Formula Weight 136.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC120935000

  SDS
fisher scientific acros
120935000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC120931000

  SDS
fisher scientific acros
120931000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 136.15
Quantity 500g
Color Clear yellow to brown
Beilstein 08, 85
TSCA TSCA
RTECS Number AM8575000
EINECS Number 204-288-0
Density 1.1310g/mL
CAS Max % 100.0
Refractive Index 1.5580 to 1.5600 (20°C, 589nm)
Synonym 1-(2-Hydroxyphenyl)ethanone
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Assay Percent Range 99%
Name Note 99%
MDL Number MFCD00002219
CAS 118-93-4
Chemical Name or Material 2'-Hydroxyacetophenone
Molecular Formula C8H8O2
Linear Formula HOC6H4COCH3
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.