Learn More
2'-Hydroxyacetophenone, 98%, Thermo Scientific Chemicals
$50.90 - $147.88
Chemical Identifiers
CAS | 118-93-4 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00002219 |
InChI Key | JECYUBVRTQDVAT-UHFFFAOYSA-N |
Synonym | 2'-hydroxyacetophenone, 2-acetylphenol, 1-2-hydroxyphenyl ethanone, o-hydroxyacetophenone, o-acetylphenol, o-hydroxyphenyl methyl ketone, ethanone, 1-2-hydroxyphenyl, 1-2-hydroxyphenyl ethan-1-one, acetophenone, 2'-hydroxy, 2-hydroxyphenyl methyl ketone |
PubChem CID | 8375 |
IUPAC Name | 1-(2-hydroxyphenyl)ethanone |
SMILES | CC(=O)C1=CC=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1255022
|
Thermo Scientific Chemicals
A1255022 |
100 g |
N/A
|
|
|||||
AAA1255036
|
Thermo Scientific Chemicals
A1255036 |
500 g |
N/A
|
|
|||||
Description
2'-Hydroxyacetophenone is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Hydroxyacetophenone is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
118-93-4 | |
136.15 | |
JECYUBVRTQDVAT-UHFFFAOYSA-N | |
8375 | |
CC(=O)C1=CC=CC=C1O |
C8H8O2 | |
MFCD00002219 | |
2'-hydroxyacetophenone, 2-acetylphenol, 1-2-hydroxyphenyl ethanone, o-hydroxyacetophenone, o-acetylphenol, o-hydroxyphenyl methyl ketone, ethanone, 1-2-hydroxyphenyl, 1-2-hydroxyphenyl ethan-1-one, acetophenone, 2'-hydroxy, 2-hydroxyphenyl methyl ketone | |
1-(2-hydroxyphenyl)ethanone |
Specifications
118-93-4 | |
1.13 | |
106°C (222°F) | |
1.5584 | |
100 g | |
2'-hydroxyacetophenone, 2-acetylphenol, 1-2-hydroxyphenyl ethanone, o-hydroxyacetophenone, o-acetylphenol, o-hydroxyphenyl methyl ketone, ethanone, 1-2-hydroxyphenyl, 1-2-hydroxyphenyl ethan-1-one, acetophenone, 2'-hydroxy, 2-hydroxyphenyl methyl ketone | |
JECYUBVRTQDVAT-UHFFFAOYSA-N | |
1-(2-hydroxyphenyl)ethanone | |
8375 | |
98% |
3°C to 6°C | |
218°C | |
C8H8O2 | |
MFCD00002219 | |
386123 | |
Slightly soluble in water. | |
CC(=O)C1=CC=CC=C1O | |
136.15 | |
136.15 | |
2'-Hydroxyacetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-288-0
RTECSNumber : AM8575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only