missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxyacetophenone, 97+%

$118.22 - $118.22

Chemical Identifiers

CAS 582-24-1
Formula Weight 136.15
MDL Number MFCD00041829
Synonym Phenacyl alcohol
Chemical Name or Material 2-Hydroxyacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4201306 SDS Alfa Aesar™
4201306
5g Each for $118.22
Specifications

Description

  • Packaged under argon
Description & Specifications

Chemical Identifiers

582-24-1
MFCD00041829
2-Hydroxyacetophenone
136.15
Phenacyl alcohol

Specifications

Air & Moisture Sensitive
5g
Powder
MFCD00041829
2-Hydroxyacetophenone
86° to 88°C
582-24-1
136.15
Phenacyl alcohol
≥97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

storageNote1 :

  • Packaged under argon