missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxy-N-methylbenzylamine Hydrochloride, TCI America™
$233.75 - $577.52
Chemical Identifiers
CAS | 60399-02-2 |
---|---|
Molecular Formula | C8H12ClNO |
Molecular Weight (g/mol) | 173.64 |
MDL Number | MFCD00142934 |
InChI Key | UDXXNWDSBJINMV-UHFFFAOYSA-N |
Synonym | alpha-Methylamino-o-cresol Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, N-Methyl-2-hydroxybenzylamine Hydrochloride |
PubChem CID | 44630388 |
IUPAC Name | 2-(methylaminomethyl)phenol;hydrochloride |
SMILES | CNCC1=CC=CC=C1O.Cl |
Chemical Identifiers
60399-02-2 | |
173.64 | |
UDXXNWDSBJINMV-UHFFFAOYSA-N | |
44630388 | |
CNCC1=CC=CC=C1O.Cl |
C8H12ClNO | |
MFCD00142934 | |
alpha-Methylamino-o-cresol Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, N-Methyl-2-hydroxybenzylamine Hydrochloride | |
2-(methylaminomethyl)phenol;hydrochloride |
Specifications
Undesignated | |
1g | |
C8H12ClNO | |
alpha-Methylamino-o-cresol Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, N-Methyl-2-hydroxybenzylamine Hydrochloride | |
CNCC1=CC=CC=C1O.Cl | |
173.64 | |
173.64 | |
≥98.0% (T) |
148°C | |
60399-02-2 | |
MFCD00142934 | |
UDXXNWDSBJINMV-UHFFFAOYSA-N | |
2-(methylaminomethyl)phenol;hydrochloride | |
44630388 | |
Crystal-Powder at 20°C | |
2-Hydroxy-N-methylbenzylamine Hydrochloride |
Safety and Handling
TSCA : No