missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-gamma-butyrolactone, 99%, ACROS Organics™

$461.87 - $461.87

Chemical Identifiers

CAS 19444-84-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C4H6O3
Formula Weight 102.09
MDL Number MFCD00134268
Chemical Name or Material 2-Hydroxy-γ-butyrolactone, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC275070050 SDS Fisher Scientific Acros
275070050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC275070250 SDS Thermo Fisher Scientific
275070250
Glass bottle 25g Each for $461.87
Specifications Description & Specifications

Chemical Identifiers

19444-84-9
100.0
C4H6O3
MFCD00134268
≥98.5%
98.5
99%
102.09
2-Hydroxy-γ-butyrolactone, 99%

Specifications

1.3130g/mL
5g
98.5
Authentic
Viscous Liquid
C4H6O3
MFCD00134268
2-Hydroxy-γ-butyrolactone, 99%
Clear colorless to yellow
19444-84-9
100.0
Glass bottle
99%
102.09
18, V,1, 3
≥98.5%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA