missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-9-fluorenone, 97%

Manufacturer:  Alfa Aesar™ H55133MD

 View more versions of this product

Catalog No. AAH55133MD



Specifications

Specifications

2-Hydroxy-9-fluorenone
204°C to 206°C
6949-73-1
MFCD00001155
GXUBPHMYNSICJC-UHFFFAOYSA-N
2-hydroxyfluoren-9-one
81386
Powder
Yellow to Orange
250mg
C13H8O2
2-hydroxy-9-fluorenone
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)O
196.205
196.21
97%

Chemical Identifiers

6949-73-1
196.205
GXUBPHMYNSICJC-UHFFFAOYSA-N
81386
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)O
C13H8O2
MFCD00001155
2-hydroxy-9-fluorenone
2-hydroxyfluoren-9-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 230-119-5

TSCA : No

Recommended Storage : Ambient temperatures

Documents