Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-6-methylpyridine, 98%

$59.96 - $221.50

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-6-methylpyridine
CAS 3279-76-3
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00006279
Beilstein 107073
Synonym 6-Hydroxy-2-picoline; 6-Methyl-2-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1476406

  SDS
alfa aesar™
A1476406
5g Each for $59.96

AAA1476414

  SDS
alfa aesar™
A1476414
25g Each for $221.50
Description & Specifications

Specifications

Formula Weight 109.13
Chemical Name or Material 2-Hydroxy-6-methylpyridine
Odor Odorless
Assay 98%
Molecular Formula C6H7NO
MDL Number MFCD00006279
CAS 3279-76-3
Beilstein 107073
Synonym 6-Hydroxy-2-picoline; 6-Methyl-2-pyridinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 157° to 161°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.