Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Hydroxy-6-methylpyridine-4-carboxylic acid, 97%

$95.54 - $1,212.95

Chemical Identifiers

Chemical Name or Material 2-Hydroxy-6-methylpyridine-4-carboxylic acid
CAS 86454-13-9
Molecular Formula C7H7NO3
Formula Weight 153.15
MDL Number MFCD02682019
Synonym 2-Hydroxy-6-methylisonicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2771603

  SDS
alfa aesar™
H2771603
1g Each for $95.54

AAH2771614

  SDS
alfa aesar™
H2771614
25g Each for $1,212.95
Description & Specifications

Specifications

Formula Weight 153.15
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Assay 97%
Molecular Formula C7H7NO3
Quantity 1g
Melting Point >300°C
MDL Number MFCD02682019
CAS 86454-13-9
Chemical Name or Material 2-Hydroxy-6-methylpyridine-4-carboxylic acid
Synonym 2-Hydroxy-6-methylisonicotinic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.