missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxy-6-methylnicotinic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1184214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Hydroxy-6-methylnicotinic acid | |
233°C to 238°C | |
MFCD00006272 | |
129164 | |
XRIHTJYXIHOBDQ-UHFFFAOYSA-N | |
6-methyl-2-oxo-1H-pyridine-3-carboxylic acid | |
94960 | |
≥98% |
38116-61-9 | |
C7H7NO3 | |
25 g | |
2-hydroxy-6-methylnicotinic acid, 2-hydroxy-6-methylpyridine-3-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid, chembl50725, 3-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 2-hydroxy-6-methyl nicotinic acid, 2-hydroxy-6-methyl-nicotinic acid, 6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid, 6-methyl-2-oxohydropyridine-3-carboxylic acid, 2-hydroxy-6-methylnicotinicacid | |
CC1=CC=C(C(=O)N1)C(=O)O | |
153.137 | |
153.14 |
Chemical Identifiers
38116-61-9 | |
153.137 | |
XRIHTJYXIHOBDQ-UHFFFAOYSA-N | |
94960 | |
CC1=CC=C(C(=O)N1)C(=O)O |
C7H7NO3 | |
MFCD00006272 | |
2-hydroxy-6-methylnicotinic acid, 2-hydroxy-6-methylpyridine-3-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid, chembl50725, 3-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 2-hydroxy-6-methyl nicotinic acid, 2-hydroxy-6-methyl-nicotinic acid, 6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid, 6-methyl-2-oxohydropyridine-3-carboxylic acid, 2-hydroxy-6-methylnicotinicacid | |
6-methyl-2-oxo-1H-pyridine-3-carboxylic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 253-784-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only