missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-6-methyl-3-nitropyridine, TCI America™

$19.86 - $55.00

Chemical Identifiers

CAS 39745-39-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD03001414
InChI Key YVYDGIGILRUPED-UHFFFAOYSA-N
Synonym 6-methyl-3-nitropyridin-2-ol, 2-hydroxy-6-methyl-3-nitropyridine, 6-hydroxy-5-nitro-2-picoline, 2-hydroxy-3-nitro-6-methylpyridine, 2-hydroxy-3-nitro-6-picoline, 6-methyl-3-nitro-2 1h-pyridinone, 6-methyl-3-nitro-2-pyridone, 3-nitro-6-methylpyridin-2-ol, 2-hydroxy-3-nitro-methylpyridine, 2 1h-pyridinone, 6-methyl-3-nitro
PubChem CID 543029
IUPAC Name 6-methyl-3-nitro-1H-pyridin-2-one
SMILES CC1=CC=C(C(=O)N1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14981G View Documents TCI America
H14981G
1g Each for $19.86
 
H14985G View Documents TCI America
H14985G
5g Each for $55.00
 
Specifications

Chemical Identifiers

39745-39-6
154.125
YVYDGIGILRUPED-UHFFFAOYSA-N
543029
CC1=CC=C(C(=O)N1)[N+](=O)[O-]
C6H6N2O3
MFCD03001414
6-methyl-3-nitropyridin-2-ol, 2-hydroxy-6-methyl-3-nitropyridine, 6-hydroxy-5-nitro-2-picoline, 2-hydroxy-3-nitro-6-methylpyridine, 2-hydroxy-3-nitro-6-picoline, 6-methyl-3-nitro-2 1h-pyridinone, 6-methyl-3-nitro-2-pyridone, 3-nitro-6-methylpyridin-2-ol, 2-hydroxy-3-nitro-methylpyridine, 2 1h-pyridinone, 6-methyl-3-nitro
6-methyl-3-nitro-1H-pyridin-2-one

Specifications

Undesignated
1g
C6H6N2O3
6-methyl-3-nitropyridin-2-ol, 2-hydroxy-6-methyl-3-nitropyridine, 6-hydroxy-5-nitro-2-picoline, 2-hydroxy-3-nitro-6-methylpyridine, 2-hydroxy-3-nitro-6-picoline, 6-methyl-3-nitro-2 1h-pyridinone, 6-methyl-3-nitro-2-pyridone, 3-nitro-6-methylpyridin-2-ol, 2-hydroxy-3-nitro-methylpyridine, 2 1h-pyridinone, 6-methyl-3-nitro
CC1=CC=C(C(=O)N1)[N+](=O)[O-]
154.125
154.13
≥98.0% (GC,T)
217°C
39745-39-6
MFCD03001414
YVYDGIGILRUPED-UHFFFAOYSA-N
6-methyl-3-nitro-1H-pyridin-2-one
543029
Crystal-Powder at 20°C
2-Hydroxy-6-methyl-3-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No