Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Hydroxy-6'-methoxyacetophenone, 97%

$482.16 - $482.16

Chemical Identifiers

Chemical Name or Material 2'-Hydroxy-6'-methoxyacetophenone
CAS 703-23-1
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00008732
Beilstein 1869049
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1853914

  SDS
alfa aesar™
A1853914
25g Each for $482.16
Description & Specifications

Specifications

Formula Weight 166.18
Assay 97%
Beilstein 1869049
CAS 703-23-1
Health Hazard 1 Warning
Chemical Name or Material 2'-Hydroxy-6'-methoxyacetophenone
Quantity 25g
Molecular Formula C9H10O3
Boiling Point 141°C (16mmHg)
MDL Number MFCD00008732
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 56° to 59°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.