missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Hydroxy-5-(trifluoromethyl)pyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC247260050
Specifications
2-Hydroxy-5-(trifluoromethyl)pyridine | |
Glass bottle | |
145.0°C to 149.0°C | |
33252-63-0 | |
100.0 | |
C6H4F3NO | |
2-hydroxy-5-trifluoromethyl pyridine, 2-hydroxy-5-trifluoromethylpyridine, 5-trifloromethyl-2-1h-pyridinone, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl pyridin-2-ol, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-2-pyridone, 5-trifluoromethyl-pyridin-2-ol, unii-1ccp81h07b | |
C1=CC(=O)NC=C1C(F)(F)F | |
163.1 | |
163.1 | |
97% |
Authentic | |
White to Beige | |
5g | |
96.0 | |
96% min. (HPLC) | |
MFCD00042315 | |
BYRJSCNPUHYZQE-UHFFFAOYSA-N | |
5-(trifluoromethyl)-1H-pyridin-2-one | |
147443 | |
Crystalline Powder |
Chemical Identifiers
33252-63-0 | |
163.1 | |
BYRJSCNPUHYZQE-UHFFFAOYSA-N | |
147443 | |
C1=CC(=O)NC=C1C(F)(F)F |
C6H4F3NO | |
MFCD00042315 | |
2-hydroxy-5-trifluoromethyl pyridine, 2-hydroxy-5-trifluoromethylpyridine, 5-trifloromethyl-2-1h-pyridinone, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl pyridin-2-ol, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-2-pyridone, 5-trifluoromethyl-pyridin-2-ol, unii-1ccp81h07b | |
5-(trifluoromethyl)-1H-pyridin-2-one |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning