missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-5-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC247260010

 View more versions of this product

Catalog No. AC247260010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Hydroxy-5-(trifluoromethyl)pyridine
Glass bottle
145.0°C to 149.0°C
33252-63-0
100.0
C6H4F3NO
2-hydroxy-5-trifluoromethyl pyridine, 2-hydroxy-5-trifluoromethylpyridine, 5-trifloromethyl-2-1h-pyridinone, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl pyridin-2-ol, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-2-pyridone, 5-trifluoromethyl-pyridin-2-ol, unii-1ccp81h07b
C1=CC(=O)NC=C1C(F)(F)F
163.1
163.1
97%
Authentic
White to Beige
1g
96.0
96% min. (HPLC)
MFCD00042315
BYRJSCNPUHYZQE-UHFFFAOYSA-N
5-(trifluoromethyl)-1H-pyridin-2-one
147443
Crystalline Powder

Chemical Identifiers

33252-63-0
163.1
BYRJSCNPUHYZQE-UHFFFAOYSA-N
147443
C1=CC(=O)NC=C1C(F)(F)F
C6H4F3NO
MFCD00042315
2-hydroxy-5-trifluoromethyl pyridine, 2-hydroxy-5-trifluoromethylpyridine, 5-trifloromethyl-2-1h-pyridinone, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl pyridin-2-ol, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-2-pyridone, 5-trifluoromethyl-pyridin-2-ol, unii-1ccp81h07b
5-(trifluoromethyl)-1H-pyridin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS Documents