missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-5-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Chemical Identifiers

CAS 33252-63-0
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.1
MDL Number MFCD00042315
InChI Key BYRJSCNPUHYZQE-UHFFFAOYSA-N
Synonym 2-hydroxy-5-trifluoromethyl pyridine
PubChem CID 147443
IUPAC Name 5-(trifluoromethyl)-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC247260010 SDS View Documents Thermo Fisher Scientific
AC247260010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC247260050 SDS View Documents Thermo Fisher Scientific
AC247260050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

33252-63-0
163.1
BYRJSCNPUHYZQE-UHFFFAOYSA-N
147443
C1=CC(=O)NC=C1C(F)(F)F
C6H4F3NO
MFCD00042315
2-hydroxy-5-trifluoromethyl pyridine
5-(trifluoromethyl)-1H-pyridin-2-one

Specifications

Authentic
White to Beige
1g
96.0
96% min. (HPLC)
MFCD00042315
BYRJSCNPUHYZQE-UHFFFAOYSA-N
5-(trifluoromethyl)-1H-pyridin-2-one
147443
Crystalline Powder
2-Hydroxy-5-(trifluoromethyl)pyridine
Glass bottle
145.0°C to 149.0°C
33252-63-0
100.0
C6H4F3NO
2-hydroxy-5-trifluoromethyl pyridine
C1=CC(=O)NC=C1C(F)(F)F
163.1
163.1
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning