Please login to your online account to display your discounted pricing

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398202500

 View more versions of this product

Catalog No. AC398202500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Low Melting Solid
Formula Weight 206.12
Quantity 250mg
Color White to pale yellow
Melting Point 30° to 34°C
CAS Max % 100.0
Chemical Name or Material 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
Assay Percent Range 99%
Molecular Formula C8H5F3O3
CAS 93249-62-8
Infrared Spectrum Authentic
NMR Consistent with structure
Packaging Glass bottle
MDL Number MFCD00075249
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CF3OC6H3(OH)CHO

Specifications

Chemical Name or Material 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
CAS 93249-62-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H5F3O3
Linear Formula CF3OC6H3(OH)CHO
Formula Weight 206.12
MDL Number MFCD00075249
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents