missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-5-nitropyridine, TCI America™

$221.53 - $1,251.28

Chemical Identifiers

CAS 5418-51-9
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00006276
InChI Key XKWSQIMYNVLGBO-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
PubChem CID 79453
IUPAC Name 5-nitro-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0673250G View Documents TCI America
H0673250G
250g Each for $1,251.28
 
H067325G View Documents TCI America
H067325G
25g Each for $221.53
 
Specifications

Chemical Identifiers

5418-51-9
140.098
XKWSQIMYNVLGBO-UHFFFAOYSA-N
79453
C1=CC(=O)NC=C1[N+](=O)[O-]
C5H4N2O3
MFCD00006276
2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
5-nitro-1H-pyridin-2-one

Specifications

Undesignated
250g
C5H4N2O3
2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
C1=CC(=O)NC=C1[N+](=O)[O-]
140.098
140.10
≥97.0% (GC,T)
186°C
5418-51-9
MFCD00006276
XKWSQIMYNVLGBO-UHFFFAOYSA-N
5-nitro-1H-pyridin-2-one
79453
Crystal-Powder at 20°C
2-Hydroxy-5-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No